Gene Transcripts

Gene Symbol
Myl4
Entrez Gene ID
688228
Gene Name
myosin, light chain 4
Gene Aliases
Chromosome Location
Chr. 10: 92628356 - 92638100 on Build Rnor_6.0
UniGene ID
Rn.216632
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109495.1 NP_001102965.1 2 212
GenBank mRNA X51531.1 - 2 191

View Full Product Details