Gene Transcripts

Gene Symbol
Mucl1
Entrez Gene ID
689178
Gene Name
mucin-like 1
Gene Aliases
Chromosome Location
Chr. 7: 145595057 - 145598721 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006242469.2 XP_006242531.1 3 498

Gene Symbol
LOC100912361
Entrez Gene ID
100912361
Gene Name
16.5 kDa submandibular gland glycoprotein-like
Gene Aliases
Chromosome Location
Chr. 7: 145564967 - 145568566 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750420.3 XP_003750468.1 317

View Full Product Details