Gene Transcripts

Gene Symbol
Tm6sf2
Entrez Gene ID
689029
Gene Name
transmembrane 6 superfamily member 2
Gene Aliases
Chromosome Location
Chr. 16: 21092483 - 21099695 on Build Rnor_6.0
UniGene ID
Rn.198368
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127654.1 NP_001121126.1 4, 5 413
GenBank mRNA BC158807.1 - 4, 5 413

View Full Product Details