Gene Transcripts

Gene Symbol
Zar1l
Entrez Gene ID
689594
Gene Name
zygote arrest 1-like
Gene Aliases
Chromosome Location
Chr. 12: 491048 - 500372 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769045.1 XP_008767267.1 3, 4 253

View Full Product Details