Gene Transcripts

Gene Symbol
Vom2r70
Entrez Gene ID
364086
Gene Name
vomeronasal 2 receptor, 70
Gene Aliases
RGD1562647
Chromosome Location
Chr. 14: 308569 - 338157 on Build Rnor_6.0
UniGene ID
Rn.217631
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099477.1 NP_001092947.1 1 167

Gene Symbol
Vom2r72
Entrez Gene ID
689072
Gene Name
vomeronasal 2 receptor, 72
Gene Aliases
Chromosome Location
Chr. 14: 765993 - 822360 on Build Rnor_6.0
UniGene ID
Rn.217638
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099517.1 NP_001092987.1 167

Gene Symbol
Vom2r73
Entrez Gene ID
689670
Gene Name
vomeronasal 2 receptor, 73
Gene Aliases
Chromosome Location
Chr. 14: 1773425 - 1791636 on Build Rnor_6.0
UniGene ID
Rn.216132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099486.1 NP_001092956.1 1 167

View Full Product Details