Gene Transcripts

Gene Symbol
JOSD1
Entrez Gene ID
9929
Gene Name
Josephin domain containing 1
Gene Aliases
dJ508I15.2
Chromosome Location
Chr. 22: 38685543 - 38701418 on Build GRCh38
UniGene ID
Hs.3094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014876.5 NP_055691.1 3 925
XM_005261876.2 XP_005261933.1 4 1232
XM_005261877.2 XP_005261934.1 4 1128
XM_005261878.2 XP_005261935.1 4 1156
XM_005261879.2 XP_005261936.1 4 1251
GenBank mRNA AK291827.1 - 3 718
BC015026.2 - 3 759 3921785
CR456473.1 - 3 604
CR456923.1 - 3 458
D31884.1 - 3 737

View Full Product Details