Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271816.1 | NP_001258745.1 | 2 | 780 | |
NM_014881.4 | NP_055696.3 | 1 | 1098 | ||
XM_006718090.1 | XP_006718153.1 | 2 | 792 | ||
XM_011540429.1 | XP_011538731.1 | 2 | 848 | ||
GenBank mRNA | BC013124.2 | - | 1 | 730 | |
BC062582.1 | - | 1 | 730 | 5202821 | |
D42045.1 | - | 1 | 982 |