Gene Transcripts

Gene Symbol
OXSR1
Entrez Gene ID
9943
Gene Name
oxidative stress responsive 1
Gene Aliases
OSR1
Chromosome Location
Chr. 3: 38165051 - 38255488 on Build GRCh38
UniGene ID
Hs.475970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005109.2 NP_005100.1 14 1639
XM_005265638.2 XP_005265695.1 15 1780
XM_011534331.2 XP_011532633.1 14 1691
XM_017007601.1 XP_016863090.1 13 1446
XM_017007602.1 XP_016863091.1 16 1966
GenBank mRNA AB017642.1 - 14 1639
AB029024.1 - 14 1392
AF087893.1 - 9 785
BC008726.2 - 14 1622 3163379

View Full Product Details