Gene Transcripts

Gene Symbol
Marc1
Entrez Gene ID
690745
Gene Name
mitochondrial amidoxime reducing component 1
Gene Aliases
Mosc1
Chromosome Location
Chr. 13: 102698522 - 102721218 on Build Rnor_6.0
UniGene ID
Rn.26676
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100811.1 NP_001094281.1 4 717
GenBank mRNA BC088229.1 - 3 281

View Full Product Details