Gene Transcripts
- Gene Symbol
- Hmgb1
- Entrez Gene ID
- 25459
- Gene Name
- high mobility group box 1
- Gene Aliases
- Ac2-008, Hmg1
- Chromosome Location
- Chr. 12: 7090927 - 7092190 on Build Rnor_6.0
- UniGene ID
- Rn.144565
- Species
- Rattus norvegicus
- Gene Symbol
- Hmg1l1
- Entrez Gene ID
- 679571
- Gene Name
- high-mobility group (nonhistone chromosomal) protein 1-like 1
- Gene Aliases
- Chromosome Location
- Chr. 19: 59984234 - 59985494 on Build Rnor_6.0
- UniGene ID
- Rn.151172
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC168143.1 |
- |
1 |
852 |
|
View Full Product Details