Gene Transcripts

Gene Symbol
Mrp63
Entrez Gene ID
691814
Gene Name
mitochondrial ribosomal protein 63
Gene Aliases
Chromosome Location
Chr. 15: 38096994 - 38097856 on Build Rnor_6.0
UniGene ID
Rn.22630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109649.2 NP_001103119.1 2 314
GenBank mRNA BC167109.1 - 2 357

View Full Product Details