Gene Transcripts

Gene Symbol
HSPB1
Entrez Gene ID
3315
Gene Name
heat shock protein family B (small) member 1
Gene Aliases
CMT2F, HEL-S-102, HMN2B, HS.76067, HSP27, HSP28, Hsp25, SRP27
Chromosome Location
Chr. 7: 76302558 - 76304297 on Build GRCh38
UniGene ID
Hs.520973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001540.3 NP_001531.1 3 749
GenBank mRNA AB020027.1 - 3 619
AK296890.1 - 4 529
AK311446.1 - 3 381
AK311894.1 - 3 632
AL050380.1 - 4 480
BC000510.2 - 3 693 2822325
BC012292.1 - 3 667 3862626
BC012768.2 - 3 616 3689220
BC014920.1 - 3 553
BC073768.1 - 3 614 5444848
BT019888.1 - 3 594
CR407614.1 - 3 594
CR536489.1 - 3 594
FJ224323.1 - 3 701
S74571.1 - 2 229
U90906.1 - 3 701
X16477.1 - 3 268
X54079.1 - 3 646

View Full Product Details