Gene Transcripts

Gene Symbol
LST1
Entrez Gene ID
7940
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144, D6S49E, LST-1
Chromosome Location
Chr. 6: 31586179 - 31588909 on Build GRCh38
UniGene ID
Hs.436066
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166538.1 NP_001160010.1 3 479
NM_007161.3 NP_009092.3 4 572
NM_205837.2 NP_995309.2 4 658
NM_205838.2 NP_995310.2 4 595
NM_205839.2 NP_995311.2 5 688
NM_205840.2 NP_995312.2 3 528
NR_029461.1 3 572
NR_029462.1 3 527
XM_006715206.3 XP_006715269.1 5 701
XM_006715209.3 XP_006715272.1 4 604
XM_006715210.3 XP_006715273.1 4 659
XM_011514914.2 XP_011513216.1 5 682
GenBank mRNA BC103855.1 - 3 463
BC103856.1 - 3 463 40003004
BX647293.1 - 2 1754

View Full Product Details