Gene Transcripts

Gene Symbol
HIST1H2AJ
Entrez Gene ID
8331
Gene Name
histone cluster 1, H2aj
Gene Aliases
H2A/E, H2AFE, dJ160A22.4
Chromosome Location
Chr. 6: 27814302 - 27814740 on Build GRCh38
UniGene ID
Hs.406691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021066.2 NP_066544.1 1 397
GenBank mRNA BC133048.1 - 1 486
BC133050.1 - 1 486 40147072

View Full Product Details