Gene Transcripts

Gene Symbol
HIST1H3D
Entrez Gene ID
8351
Gene Name
histone cluster 1, H3d
Gene Aliases
H3/b, H3FB
Chromosome Location
Chr. 6: 26196784 - 26199293 on Build GRCh38
UniGene ID
Hs.532144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003530.4 NP_003521.2 2 867
GenBank mRNA BC031333.1 - 2 786

View Full Product Details