Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320296.1 | NP_001307225.1 | 1 | 550 | |
NM_003717.3 | NP_003708.1 | 2 | 320 | ||
GenBank mRNA | AF005271.1 | - | 2 | 319 | |
BC104234.1 | - | 1 | 499 | 40033026 | |
BC104235.1 | - | 1 | 499 | 40033028 |