Gene Transcripts

Gene Symbol
CCDC85B
Entrez Gene ID
11007
Gene Name
coiled-coil domain containing 85B
Gene Aliases
DIPA
Chromosome Location
Chr. 11: 65890404 - 65891635 on Build GRCh38
UniGene ID
Hs.66713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006848.2 NP_006839.2 1 782
GenBank mRNA AK312109.1 - 1 542
BC008796.2 - 1 547 3617697
U63825.1 - 1 430

View Full Product Details