Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
84266
Gene Name
alkB homolog 7
Gene Aliases
ABH7, SPATA11, UNQ6002
Chromosome Location
Chr. 19: 6372433 - 6375250 on Build GRCh38
UniGene ID
Hs.111099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032306.3 NP_115682.1 1 572
XM_017027355.1 XP_016882844.1 1 208
GenBank mRNA AK223336.1 - 1 195
AK311956.1 - 1 211
AY358858.1 - 1 572
AY427650.2 - 1 572
BC004393.1 - 1 227 3635540
BX334356.2 - 1 489

View Full Product Details