Gene Transcripts
- Gene Symbol
- ULK4P2
- Entrez Gene ID
- 100288380
- Gene Name
- ULK4 pseudogene 2
- Gene Aliases
- D-X, FAM7A2
- Chromosome Location
- Chr. 15: 30572555 - 30600708 on Build GRCh38
- UniGene ID
- Hs.642616
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027470.1 |
|
4 |
237 |
|
GenBank mRNA |
BC137222.1 |
- |
3 |
220 |
|
- Gene Symbol
- ULK4P1
- Entrez Gene ID
- 89838
- Gene Name
- ULK4 pseudogene 1
- Gene Aliases
- D-X, FAM7A1
- Chromosome Location
- Chr. 15: 32406611 - 32435049 on Build GRCh38
- UniGene ID
- Hs.574255
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026858.1 |
|
4 |
237 |
|
GenBank mRNA |
BC071630.1 |
- |
4 |
237 |
|
BC137210.1 |
- |
5 |
331 |
|
- Gene Symbol
- ULK4P3
- Entrez Gene ID
- 89837
- Gene Name
- ULK4 pseudogene 3
- Gene Aliases
- D-X, FAM7A3
- Chromosome Location
- Chr. 15: 30103732 - 30131748 on Build GRCh38
- UniGene ID
- Hs.510848
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026859.1 |
|
4 |
496 |
|
GenBank mRNA |
BC023564.2 |
- |
4 |
496 |
|
BC071855.1 |
- |
4 |
494 |
|
View Full Product Details