Gene Transcripts

Gene Symbol
KCNG4
Entrez Gene ID
93107
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Gene Aliases
KV6.3, KV6.4
Chromosome Location
Chr. 16: 84221135 - 84239750 on Build GRCh38
UniGene ID
Hs.335877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172347.2 NP_758857.1 2 432
GenBank mRNA AF348984.1 - 1 311
AF450108.1 - 1 311
BC008969.2 - 2 432
BC109203.2 - 2 380 40005993

View Full Product Details