Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080441.1 | NP_001073910.1 | 1 | 71 | |
GenBank mRNA | BC146916.1 | - | 1 | 94 | |
BC146921.1 | - | 1 | 94 | 9056849 | |
BC171906.1 | - | 1 | 94 | 9054560 | |
BC171913.1 | - | 1 | 94 | 9054567 | |
EU489483.1 | - | 1 | 133 |