Gene Transcripts

Gene Symbol
EIF4E1B
Entrez Gene ID
253314
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
Chromosome Location
Chr. 5: 176630613 - 176646644 on Build GRCh38
UniGene ID
Hs.448362
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099408.1 NP_001092878.1 7 1035
XM_011534499.2 XP_011532801.1 7 1116
GenBank mRNA AK094270.1 - 6 1403
BM709255.1 - 2 123

View Full Product Details