Gene Transcripts

Gene Symbol
KRTAP19-2
Entrez Gene ID
337969
Gene Name
keratin associated protein 19-2
Gene Aliases
KAP19.2
Chromosome Location
Chr. 21: 30487190 - 30487348 on Build GRCh38
UniGene ID
Hs.553692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181608.1 NP_853639.1 1 65
GenBank mRNA AB180041.1 - 1 153

View Full Product Details