Gene Transcripts

Gene Symbol
KRTAP20-2
Entrez Gene ID
337976
Gene Name
keratin associated protein 20-2
Gene Aliases
KAP20.2
Chromosome Location
Chr. 21: 30635264 - 30635461 on Build GRCh38
UniGene ID
Hs.553698
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181616.1 NP_853647.1 1 41
GenBank mRNA AB096957.1 - 1 69
BC111084.1 - 1 41 40013739

View Full Product Details