Gene Transcripts

Gene Symbol
SPDYE4
Entrez Gene ID
388333
Gene Name
speedy/RINGO cell cycle regulator family member E4
Gene Aliases
Chromosome Location
Chr. 17: 8746868 - 8759428 on Build GRCh38
UniGene ID
Hs.368429
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128076.1 NP_001121548.1 5 754
XM_005256632.4 XP_005256689.1 5 1623
XM_005256633.4 XP_005256690.1 5 1623
GenBank mRNA BC146949.1 - 5 754
BC146966.1 - 5 754 9056894

View Full Product Details