Gene Transcripts

Gene Symbol
GOLGA8EP
Entrez Gene ID
390535
Gene Name
golgin A8 family member E, pseudogene
Gene Aliases
GOLGA8E
Chromosome Location
Chr. 15: 22424673 - 22438026 on Build GRCh38
UniGene ID
Hs.454647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033350.1 18 2328
GenBank mRNA BC063309.1 - 19 2302
DQ309037.1 - 18 2302

View Full Product Details