Gene Transcripts

Gene Symbol
MROH6
Entrez Gene ID
642475
Gene Name
maestro heat like repeat family member 6
Gene Aliases
C8orf73
Chromosome Location
Chr. 8: 143566189 - 143572758 on Build GRCh38
UniGene ID
Hs.531406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100878.1 NP_001094348.1 1 111
XM_006716615.2 XP_006716678.1 1 105
XM_011517214.1 XP_011515516.1 1 105
XM_011517215.1 XP_011515517.1 1 105
XM_011517216.2 XP_011515518.1 1 105
XM_011517217.1 XP_011515519.1 1 105
XM_011517218.1 XP_011515520.1 1 105
XM_011517219.1 XP_011515521.1 1 105
XM_011517220.1 XP_011515522.1 1 105
XM_011517221.2 XP_011515523.1 1 105
XM_011517222.1 XP_011515524.1 1 105
XM_011517223.1 XP_011515525.1 1 105
GenBank mRNA BX341537.2 - 1 111

View Full Product Details