Gene Transcripts
- Gene Symbol
- PREB
- Entrez Gene ID
- 10113
- Gene Name
- prolactin regulatory element binding
- Gene Aliases
- SEC12
- Chromosome Location
- Chr. 2: 27130756 - 27134674 on Build GRCh38
- UniGene ID
- Hs.279784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013388.4 |
NP_037520.1
|
7, 8 |
1258 |
|
XM_006711914.2 |
XP_006711977.1
|
6, 7 |
1065 |
|
GenBank mRNA |
AF203687.1 |
- |
7, 8 |
1136 |
|
AF226684.1 |
- |
7, 8 |
1149 |
|
AK023064.1 |
- |
7, 8 |
1215 |
|
AK023405.1 |
- |
7, 8 |
1129 |
|
AK292178.1 |
- |
8, 9 |
1145 |
|
BC002765.2 |
- |
7, 8 |
1189 |
3631721 |
BC012890.1 |
- |
7, 8 |
1199 |
4158577 |
BC016472.1 |
- |
7, 8 |
1189 |
4178911 |
BC016906.1 |
- |
7, 8 |
1132 |
4427483 |
BC041032.1 |
- |
7, 8 |
1252 |
4638313 |
View Full Product Details