Gene Transcripts

Gene Symbol
DNAL4
Entrez Gene ID
10126
Gene Name
dynein axonemal light chain 4
Gene Aliases
MRMV3, PIG27
Chromosome Location
Chr. 22: 38778508 - 38794156 on Build GRCh38
UniGene ID
Hs.632766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005740.2 NP_005731.1 4 1399
GenBank mRNA AL035366.1 - 4 1392
BC002968.1 - 4 1366 3543962

View Full Product Details