Gene Transcripts

Gene Symbol
TEX22
Entrez Gene ID
647310
Gene Name
testis expressed 22
Gene Aliases
Chromosome Location
Chr. 14: 105398499 - 105413859 on Build GRCh38
UniGene ID
Hs.729499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195082.1 NP_001182011.1 3, 4 379
XM_006720234.3 XP_006720297.1 3, 4 653
GenBank mRNA BX378619.2 - 3, 4 379

View Full Product Details