Gene Transcripts

Gene Symbol
ASAH2B
Entrez Gene ID
653308
Gene Name
N-acylsphingosine amidohydrolase 2B
Gene Aliases
ASAH2C, ASAH2L, bA449O16.3, bA98I6.3
Chromosome Location
Chr. 10: 50739688 - 50757188 on Build GRCh38
UniGene ID
Hs.659820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079516.3 NP_001072984.1 2, 3 414
NM_001321957.1 NP_001308886.1 2, 3 395
GenBank mRNA BG773189.1 - 2, 3 158

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
Chromosome Location
Chr. 10: 50184861 - 50251564 on Build GRCh38
UniGene ID
Hs.512645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017016436.1 XP_016871925.1 2, 3 193

View Full Product Details