Gene Transcripts
- Gene Symbol
- HIST2H2BF
- Entrez Gene ID
- 440689
- Gene Name
- histone cluster 2, H2bf
- Gene Aliases
- Chromosome Location
- Chr. 1: 149782689 - 149812373 on Build GRCh38
- UniGene ID
- Hs.632451
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092418.1 |
- |
1 |
1051 |
|
- Gene Symbol
- LOC100996318
- Entrez Gene ID
- 100996318
- Gene Name
- uncharacterized LOC100996318
- Gene Aliases
- Chromosome Location
- Chr. 1: 121090289 - 121098265 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC100996713
- Entrez Gene ID
- 100996713
- Gene Name
- uncharacterized LOC100996713
- Gene Aliases
- Chromosome Location
- Chr. 1: 143878414 - 143885954 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details