Gene Transcripts

Gene Symbol
CNKSR1
Entrez Gene ID
10256
Gene Name
connector enhancer of kinase suppressor of Ras 1
Gene Aliases
CNK, CNK1
Chromosome Location
Chr. 1: 26177490 - 26189884 on Build GRCh38
UniGene ID
Hs.16232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297647.1 NP_001284576.1 17, 18 1617
NM_001297648.1 NP_001284577.1 17, 18 1591
NM_006314.2 NP_006305.2 17, 18 1596
XM_011540484.2 XP_011538786.1 17, 18 1576
XM_011540485.2 XP_011538787.1 14, 15 1106
GenBank mRNA AB209335.1 - 17, 18 1590
AF100153.1 - 17, 18 1574
AK001900.1 - 17, 18 1570
AK222817.1 - 17, 18 1575
AK222904.1 - 17, 18 1579
AK292646.1 - 17, 18 1578
BC011604.2 - 17, 18 1598 3139227
BC012797.2 - 17, 18 1598 3505293
BT006900.1 - 17, 18 1559

View Full Product Details