Gene Transcripts

Gene Symbol
PRLH
Entrez Gene ID
51052
Gene Name
prolactin releasing hormone
Gene Aliases
PRH, PRRP
Chromosome Location
Chr. 2: 237566574 - 237567175 on Build GRCh38
UniGene ID
Hs.247710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015893.1 NP_056977.1 1 69
GenBank mRNA AB015419.1 - 1 69
BC069081.1 - 1 69 7216895
BC069284.1 - 1 69 7262679

View Full Product Details