Gene Transcripts

Gene Symbol
LARP6
Entrez Gene ID
55323
Gene Name
La ribonucleoprotein domain family member 6
Gene Aliases
ACHN
Chromosome Location
Chr. 15: 70831524 - 70854265 on Build GRCh38
UniGene ID
Hs.416755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197958.2 NP_932062.1 2 409
GenBank mRNA BC039153.1 - 2 265

View Full Product Details