Gene Transcripts

Gene Symbol
USP41
Entrez Gene ID
373856
Gene Name
ubiquitin specific peptidase 41
Gene Aliases
Chromosome Location
Chr. 22: 20363621 - 20390758 on Build GRCh38
UniGene ID
Hs.38260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017029164.1 XP_016884653.1 2 50

View Full Product Details