Gene Transcripts

Gene Symbol
Aspm
Entrez Gene ID
12316
Gene Name
asp (abnormal spindle)-like, microcephaly associated (Drosophila)
Gene Aliases
Calmbp1, D330028K02Rik, MCPH5, Sha1
Chromosome Location
Chr. 1: 139454773 - 139494091 on Build GRCm38
UniGene ID
Mm.168523
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009791.4 NP_033921.3 18 5255
GenBank mRNA AF533752.1 - 18 5233
BC026699.1 - 2 1306

View Full Product Details