Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007637.2 | NP_031663.1 | 3 | 398 | |
GenBank mRNA | AK077753.1 | - | 3 | 356 | |
AK088184.1 | - | 3 | 332 | ||
AK129056.1 | - | 3 | 304 | ||
AK133857.1 | - | 3 | 356 | ||
AK136493.1 | - | 3 | 350 | ||
AK145445.1 | - | 3 | 367 | ||
AK146405.1 | - | 3 | 370 | ||
AK146425.1 | - | 3 | 350 | ||
AK150390.1 | - | 3 | 317 | ||
AK153911.1 | - | 3 | 374 | ||
AK167895.1 | - | 3 | 339 | ||
AK217822.1 | - | 2 | 151 | ||
BC092233.1 | - | 3 | 368 | ||
BC094427.1 | - | 3 | 345 | 30537966 | |
Z31555.1 | - | 3 | 398 |