Gene Transcripts
- Gene Symbol
- MARCH6
- Entrez Gene ID
- 10299
- Gene Name
- membrane associated ring-CH-type finger 6
- Gene Aliases
- DOA10, MARCH-VI, RNF176, TEB4
- Chromosome Location
- Chr. 5: 10353639 - 10440388 on Build GRCh38
- UniGene ID
- Hs.432862
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270660.1 |
NP_001257589.1
|
17, 18 |
1806 |
|
NM_001270661.1 |
NP_001257590.1
|
15, 16 |
1635 |
|
NM_005885.3 |
NP_005876.2
|
18, 19 |
1950 |
|
XM_011513932.2 |
XP_011512234.1
|
18, 19 |
1708 |
|
XM_011513934.1 |
XP_011512236.1
|
18, 19 |
1911 |
|
XM_011513936.2 |
XP_011512238.1
|
18, 19 |
1911 |
|
XM_011513937.2 |
XP_011512239.1
|
8, 9 |
1292 |
|
GenBank mRNA |
AB011169.1 |
- |
18, 19 |
1873 |
|
AF009301.1 |
- |
5, 6 |
569 |
|
AK296585.1 |
- |
15, 16 |
1558 |
|
AK300034.1 |
- |
17, 18 |
1742 |
|
AK314686.1 |
- |
7, 8 |
803 |
|
BC046148.1 |
- |
10, 11 |
867 |
|
BC136461.1 |
- |
18, 19 |
1822 |
9020450 |
BC136462.1 |
- |
18, 19 |
1822 |
9020451 |
BC142679.1 |
- |
18, 19 |
1801 |
40148250 |
BC142694.1 |
- |
18, 19 |
1801 |
40148713 |
View Full Product Details