Gene Transcripts

Gene Symbol
Hmx1
Entrez Gene ID
15371
Gene Name
H6 homeobox 1
Gene Aliases
Nkx5-3
Chromosome Location
Chr. 5: 35388495 - 35400455 on Build GRCm38
UniGene ID
Mm.358606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010445.2 NP_034575.1 2 634
GenBank mRNA AF009367.1 - 2 634

View Full Product Details