Gene Transcripts

Gene Symbol
Smad4
Entrez Gene ID
17128
Gene Name
SMAD family member 4
Gene Aliases
AW743858, D18Wsu70e, DPC4, Madh4
Chromosome Location
Chr. 18: 73638998 - 73703741 on Build GRCm38
UniGene ID
Mm.100399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008540.2 NP_032566.2 12 2900
GenBank mRNA AK004804.1 - 11 2265
AK170255.1 - 1 381
BC046584.1 - 12 2900 6313280

View Full Product Details