Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202413.1 | NP_001189342.1 | 9 | 1162 | |
NM_001202414.1 | NP_001189343.1 | 10 | 1290 | ||
NM_006066.3 | NP_006057.1 | 9 | 1379 | ||
NM_153326.2 | NP_697021.1 | 8 | 1251 | ||
XM_011540491.2 | XP_011538793.1 | 7 | 1110 | ||
XM_011540492.2 | XP_011538794.1 | 8 | 1076 | ||
GenBank mRNA | AK293083.1 | - | 8 | 987 | |
BC000670.2 | - | 9 | 1308 | 3349300 | |
BC005394.1 | - | 8 | 967 | 4051944 | |
BT007003.1 | - | 7 | 872 | ||
CR457010.1 | - | 7 | 872 | ||
EU668319.1 | - | 8 | 1208 | ||
EU794588.1 | - | 8 | 1208 | ||
J04794.1 | - | 8 | 932 |