Gene Transcripts

Gene Symbol
Tsx
Entrez Gene ID
22127
Gene Name
testis specific X-linked gene
Gene Aliases
AV207184
Chromosome Location
Chr. X: 103409101 - 103424588 on Build GRCm38
UniGene ID
Mm.358644
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009440.2 NP_033466.1 3 234
GenBank mRNA BC048493.1 - 3 234
HQ433540.1 - 4 345
HQ433541.1 - 4 449
HQ433542.1 - 4 344
HQ433544.1 - 3 534
HQ433547.1 - 3 434
X99796.1 - 3 219

View Full Product Details