Gene Transcripts

Gene Symbol
Vmn2r47
Entrez Gene ID
100042891
Gene Name
vomeronasal 2, receptor 47
Gene Aliases
Chromosome Location
Chr. 7: 9816782 - 9841325 on Build GRCm38
UniGene ID
Mm.465205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105151.1 NP_001098621.1 3 838

Gene Symbol
Vmn2r30
Entrez Gene ID
22306
Gene Name
vomeronasal 2, receptor 30
Gene Aliases
ENSMUSG00000070847, V2r15
Chromosome Location
Chr. 7: 7312274 - 7337493 on Build GRCm38
UniGene ID
Mm.458977
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009490.3 NP_033516.3 3 838
GenBank mRNA AF011425.1 - 877

View Full Product Details