Gene Transcripts

Gene Symbol
Slc16a4
Entrez Gene ID
229699
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 4
Gene Aliases
AW146050, MCT 5
Chromosome Location
Chr. 3: 107289865 - 107312115 on Build GRCm38
UniGene ID
Mm.38384
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146136.1 NP_666248.1 6 772
GenBank mRNA AK045797.1 - 6 758
AK049873.1 - 6 835
AK052779.1 - 6 771
AK085492.1 - 6 769
AK136837.1 - 6 771
BC025441.1 - 6 760 5043709
BC026596.1 - 6 772 4975010

View Full Product Details