Gene Transcripts

Gene Symbol
Plekha7
Entrez Gene ID
233765
Gene Name
pleckstrin homology domain containing, family A member 7
Gene Aliases
A430081P20Rik, AI414265
Chromosome Location
Chr. 7: 116124118 - 116308381 on Build GRCm38
UniGene ID
Mm.3741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK040271.1 - 22 3318
AK146866.1 - 3509
BC060678.1 - 21 3239
EU380771.1 - 3535
EU380772.1 - 3593

View Full Product Details