Gene Transcripts

Gene Symbol
Il17d
Entrez Gene ID
239114
Gene Name
interleukin 17D
Gene Aliases
AI462269, IL-17D, Il27a
Chromosome Location
Chr. 14: 57524829 - 57543166 on Build GRCm38
UniGene ID
Mm.390726
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145837.3 NP_665836.2 3 844
GenBank mRNA AF458063.1 - 2 766
AK131917.1 - 3 893
BC099514.1 - 1 286

View Full Product Details