Gene Transcripts

Gene Symbol
Gfm2
Entrez Gene ID
320806
Gene Name
G elongation factor, mitochondrial 2
Gene Aliases
6530419G12Rik, A930009M04Rik, EF-G2mt, EFG2, MST027, RRF2mt
Chromosome Location
Chr. 13: 97137937 - 97181196 on Build GRCm38
UniGene ID
Mm.219675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146043.2 NP_001139515.1 10 949
NM_001271463.1 NP_001258392.1 10 949
NM_001271464.1 NP_001258393.1 10 949
NM_001271465.1 NP_001258394.1 10 976
NM_177266.5 NP_796240.3 10 982
GenBank mRNA AK028450.1 - 10 935
AK035819.1 - 10 888
AK044371.1 - 10 950
AK083441.1 - 10 914
AK161797.1 - 10 885
AK165871.1 - 10 943
BC027341.1 - 10 933 3984129
BC138233.1 - 10 880 8861252
BC145208.1 - 10 880 9053759

View Full Product Details