Gene Transcripts

Gene Symbol
Speer4e
Entrez Gene ID
624245
Gene Name
spermatogenesis associated glutamate (E)-rich protein 4e
Gene Aliases
Chromosome Location
Chr. 5: 14933631 - 14938475 on Build GRCm38
UniGene ID
Mm.368256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122661.1 NP_001116133.1 5 1085

View Full Product Details