Gene Transcripts

Gene Symbol
Nlrp1c-ps
Entrez Gene ID
627984
Gene Name
NLR family, pyrin domain containing 1C, pseudogene
Gene Aliases
Nalp1c, Nlrp1c, OTTMUSG00000006090
Chromosome Location
Chr. 11: 71242430 - 71285232 on Build GRCm38
UniGene ID
Mm.390378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC141384.1 - 2 676
DQ117602.1 - 2 680

View Full Product Details